CAS RN 370094-12-5
CAS RN 370094-12-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Diltiazem Sulfoxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Diltiazem Sulphoxide, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Diltiazem Sulphoxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.