CAS RN 3690-10-6
CAS RN 3690-10-6
IUPAC Name:
1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms:
zebularine
Molecular Weight (g/mol):
228.2
Molecular Formula:
C9H12N2O5
InChi Key:
RPQZTTQVRYEKCR-UHFFFAOYNA-N
SMILES:
OCC1OC(C(O)C1O)N1C=CC=NC1=O
1
–
3
of
3
results
1
Zebularine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Zebularine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3