Learn More
Zebularine, TRC
Specifications
Specifications
| Chemical Name or Material | Zebularine |
| CAS | 3690-10-6 |
| Molecular Formula | C9 H12 N2 O5 |
| Quantity | 25 mg |
| Synonym | 1-β-D-Ribofuranosyl-2(1H)-pyrimidinone, 4-Deoxyuridine, NSC 309132, Pyrimidin-2-one β-D-Ribofuranoside |
| InChI Formula | InChI=1S/C9H12N2O5/c12-4-5-6(13)7(14)8(16-5)11-3-1-2-10-9(11)15/h1-3,5-8,12-14H,4H2/t5-,6-,7-,8-/m1/s1 |
| SMILES | OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC=NC2=O |
| IUPAC Name | 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
| Molecular Weight (g/mol) | 228.2 |
| Formula Weight | 228.0746 |