CAS RN 3689-24-5
CAS RN 3689-24-5
IUPAC Name:
O,O-diethyl {[diethoxy(sulfanylidene)-λ⁵-phosphanyl]oxy}phosphonothioate
Synonyms:
dithione caswell no. 837 fulex Sulpho-TEPP rcra waste number P109 tetraethyl ester dithiodiphosphoric acid tetraethyldithiopyrophosphate lethalaire G-57 ethyl thiopyrophosphate ((eto)2(PS)2O) Sulfo-TEPP
Molecular Weight (g/mol):
322.31
Molecular Formula:
C8H20O5P2S2
InChi Key:
XIUROWKZWPIAIB-UHFFFAOYSA-N
SMILES:
CCOP(=S)(OCC)OP(=S)(OCC)OCC
1
–
5
of
5
results
1
Sulfotep, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Sulfotep, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Sulfotep 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
4
Sulfotep, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.