accessibility menu, dialog, popup

UserName

CAS RN 3535-83-9

IH3CNCH3CH3CH3

CAS RN 3535-83-9

IUPAC Name: tetraheptylazanium iodide
Synonyms: tetraheptylazanium iodide
Molecular Weight (g/mol): 537.7
Molecular Formula: C28H60IN
InChi Key: KCSOHLKZTZMKQA-UHFFFAOYSA-M
SMILES: [I-].CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results