CAS RN 32980-71-5
IUPAC Name:
tetrachloropyrimido[5,4-d][1,3]diazine
Synonyms:
tetrachloropyrimido[5,4-d][1,3]diazine
Molecular Weight (g/mol):
269.89
Molecular Formula:
C6Cl4N4
InChi Key:
QNKFHUMDHRWWES-UHFFFAOYSA-N
SMILES:
ClC1=NC(Cl)=C2N=C(Cl)N=C(Cl)C2=N1
1
–
3
of
3
results
1
2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.