Learn More
2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine |
| CAS | 32980-71-5 |
| Molecular Formula | C6 Cl4 N4 |
| Quantity | 500 mg |
| Synonym | Pyrimido[5,4-d]pyrimidine','2,4,6,8-tetrachloro-, Pyrimido[5,4-d]pyrimidine','tetrachloro- (9CI), 2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine, NSC 96654, Tetrachloropyrimido[5,4-d]pyrimidine |
| InChI Formula | InChI=1S/C6Cl4N4/c7-3-1-2(12-6(10)13-3)4(8)14-5(9)11-1 |
| SMILES | Clc1nc(Cl)c2nc(Cl)nc(Cl)c2n1 |
| IUPAC Name | 2,4,6,8-tetrachloropyrimido[5,4-d]pyrimidine |
| Molecular Weight (g/mol) | 269.9 |
| Formula Weight | 267.8877 |