accessibility menu, dialog, popup

UserName

CAS RN 31676-49-0

IHNO

CAS RN 31676-49-0

IUPAC Name: 7-iodo-2,3-dihydro-1H-indol-2-one
Synonyms: 7-iodo-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 259.05
Molecular Formula: C8H6INO
InChi Key: KLJWMAKCVALVKZ-UHFFFAOYSA-N
SMILES: IC1=CC=CC2=C1NC(=O)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results