Learn More
7-Iodooxindole, 96%, Thermo Scientific Chemicals
Product Code. 15478987
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478987 250 mg 250mg
15468987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15478987 Supplier Thermo Scientific Alfa Aesar Supplier No. H52337.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31676-49-0
Molecular Formula C8H6INO
Molecular Weight (g/mol) 259.046
MDL Number MFCD09835634
InChI Key KLJWMAKCVALVKZ-UHFFFAOYSA-N
Synonym 7-iodooxindole, 7-?iodooxindole, 7-iodo-2-indolinone, 7-iodoindoline-2-one, 2h-indol-2-one, 1,3-dihydro-7-iodo, 7-iodo-1,3-dihydro-indol-2-one
PubChem CID 11470918
IUPAC Name 7-iodo-1,3-dihydroindol-2-one
SMILES C1C2=C(C(=CC=C2)I)NC1=O
Specifications
Melting Point 208°C to 209°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 259.04
Percent Purity 96%
Chemical Name or Material 7-Iodooxindole