accessibility menu, dialog, popup

UserName

CH3NHPSOH3COCH3OOH3CCH3(Z)

CAS RN 31218-83-4

IUPAC Name: propan-2-yl (2Z)-3-{[(ethylamino)(methoxy)sulfanylidene-λ⁵-phosphanyl]oxy}but-2-enoate
Synonyms: isopropyl (2Z)-3-{[(ethylamino)(methoxy)sulfanylidene-λ⁵-phosphanyl]oxy}but-2-enoate
Molecular Weight (g/mol): 281.31
Molecular Formula: C10H20NO4PS
InChi Key: BZNDWPRGXNILMS-CLFYSBASNA-N
SMILES: CCNP(=S)(OC)O\C(C)=C/C(=O)OC(C)C
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
Concentration 
  • (2)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)