Learn More
Propetamphos (>85%), TRC
Specifications
Specifications
| CAS | 31218-83-4 |
| Molecular Formula | C10 H20 N O4 P S |
| Quantity | 50 mg |
| Synonym | 2-Butenoic acid, 3-[[(ethylamino)methoxyphosphinothioyl]oxy]-, 1-methylethyl ester, (E)-, Crotonic acid, 3-hydroxy-, isopropyl ester, O-ester with O-methyl ethylphosphoramidothioate, (E)- (8CI), Blotic, Deadmag, E-Propetamphos, Ectomort Centenary, Ectomort Jetting Fluid, Ectomort Jetting Fluid and Sheep Dip, Magget, Mules-n-Mark II, Propetamphos, SAN 322, SAN 322I, Safrotin, Safrotin Brush-on Bait, Safrotin MC, Safrotin S 200, VEL 4283, Young's Summer Dip, 2-Butenoic acid, 3-[[(ethylamino)methoxyphosphinothioyl]oxy]-, 1-methylethyl ester, (2E)- |
| InChI Formula | InChI=1S/C10H20NO4PS/c1-6-11-16(17,13-5)15-9(4)7-10(12)14-8(2)3/h7-8H,6H2,1-5H3,(H,11,17)/b9-7+ |
| SMILES | CCNP(=S)(OC)O\C(=C\C(=O)OC(C)C)\C |
| IUPAC Name | propan-2-yl (E)-3-[ethylamino(methoxy)phosphinothioyl]oxybut-2-enoate |
| Molecular Weight (g/mol) | 281.31 |
| Formula Weight | 281.0851 |
| Chemical Name or Material | Propetamphos |