accessibility menu, dialog, popup

UserName

CAS RN 3096-56-8

BrO

CAS RN 3096-56-8

IUPAC Name: 2-bromo-9H-fluoren-9-one
Synonyms: 2-bromofluoren-9-one
Molecular Weight (g/mol): 259.1
Molecular Formula: C13H7BrO
InChi Key: MTCARZDHUIEYMB-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)