Learn More
2-Bromo-9-fluorenone, 96%
Product Code. 15432368
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432368 5 g 5g
15422368 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15432368 Supplier Thermo Scientific Alfa Aesar Supplier No. H55962.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 3096-56-8 | C13H7BrO | 259.10 g/mol
2-Bromo-9-fluorenone, is used as organic intermediates functional materials. It is mainly used as best liquid crystal material. It is used as an OLED intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-9-fluorenone, is used as organic intermediates functional materials. It is mainly used as best liquid crystal material. It is used as an OLED intermediate.
Solubility
Solubility in other solvents, soluble in acetone.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 3096-56-8
Molecular Formula C13H7BrO
Molecular Weight (g/mol) 259.10
MDL Number MFCD00010789
InChI Key MTCARZDHUIEYMB-UHFFFAOYSA-N
Synonym 2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f
PubChem CID 725831
IUPAC Name 2-bromofluoren-9-one
SMILES BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1
Spécification
Melting Point 146°C to 148°C
Quantity 5 g
Formula Weight 259.1
Percent Purity 96%
Chemical Name or Material 2-Bromo-9-fluorenone
RUO – Research Use Only
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.