accessibility menu, dialog, popup

UserName

CH3H3COHCH3H3CHO

CAS RN 3010-96-6

IUPAC Name: 2,2,4,4-tetramethylcyclobutane-1,3-diol
Synonyms: 2,2,4,4-tetramethylcyclobutane-1,3-diol
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: FQXGHZNSUOHCLO-UHFFFAOYSA-N
SMILES: CC1(C)C(O)C(C)(C)C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)