Learn More
2,2,4,4-Tetramethyl-1,3-cyclobutanediol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dihydroxy-1,1,3,3-tetramethylcyclobutane |
| CAS | 3010-96-6 |
| Molecular Formula | C8 H16 O2 |
| Quantity | 100 mg |
| Synonym | 2,2,4,4-Tetramethyl-1,3-cyclobutanediol, 1,3-Dihydroxy-2,2,4,4-tetramethylcyclobutane, 2,2,4,4-Tetramethyl-1,3-cyclobutandiol, 2,4-Dihydroxy-1,1,3,3-tetramethylcyclobutane, NSC 46473, Tetramethyl-1,3-cyclobutanediol, 1,3-Cyclobutanediol','2,2,4,4-tetramethyl- |
| InChI Formula | InChI=1S/C8H16O2/c1-7(2)5(9)8(3,4)6(7)10/h5-6,9-10H,1-4H3 |
| SMILES | CC1(C)C(O)C(C)(C)C1O |
| IUPAC Name | 2,2,4,4-tetramethylcyclobutane-1,3-diol |
| Molecular Weight (g/mol) | 144.211 |
| Formula Weight | 144.115 |