CAS RN 3006-86-8
IUPAC Name:
1,1-bis(tert-butylperoxy)cyclohexane
Synonyms:
1,1-bis(tert-butylperoxy)cyclohexane
Molecular Weight (g/mol):
260.37
Molecular Formula:
C14H28O4
InChi Key:
HSLFISVKRDQEBY-UHFFFAOYSA-N
SMILES:
CC(C)(C)OOC1(CCCCC1)OOC(C)(C)C
1
–
1
of
1
results
1
1,1-Di(tert-butylperoxy)cyclohexane, 50% solution in mineral oil
CAS: 3006-86-8 | C14H28O4 | 260.374 g/mol