CAS RN 2985481-46-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.