CAS RN 297-97-2
IUPAC Name:
O,O-diethyl O-pyrazin-2-yl phosphorothioate
Synonyms:
thionazin
Molecular Weight (g/mol):
248.24
Molecular Formula:
C8H13N2O3PS
InChi Key:
IRVDMKJLOCGUBJ-UHFFFAOYSA-N
SMILES:
CCOP(=S)(OCC)OC1=CN=CC=N1
1
–
4
of
4
results
1
Thionazin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Thionazin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Thionazin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.