Learn More
Thionazin, TRC
Specifications
Specifications
| Chemical Name or Material | Thionazin |
| CAS | 297-97-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8 H13 N2 O3 P S |
| Quantity | 25 mg |
| Synonym | Phosphorothioic acid','O,O-diethyl O-2-pyrazinyl ester, Ethyl pyrazinyl phosphorothioate (6CI), Phosphorothioic acid','O,O-diethyl O-pyrazinyl ester (8CI,9CI), AC 18133, ACC 18133, American Cyanamid 18133, CL 18133, Cynem, Cynophos, EN 18133, ENT 25,580, Experimental nematocide 18,133, Nemafos, Nemafos 10 G, Nemaphos, Nematocide GR, O,O-Diethyl O-2-pyrazinyl phosphorothioate, Thionazin, Thionazine, Zinophos, Zynophos |
| InChI Formula | InChI=1S/C8H13N2O3PS/c1-3-11-14(15,12-4-2)13-8-7-9-5-6-10-8/h5-7H,3-4H2,1-2H3 |
| SMILES | CCOP(=S)(OCC)Oc1cnccn1 |
| IUPAC Name | diethoxy-pyrazin-2-yloxy-sulfanylidene-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 248.24 |
| Show More |