CAS RN 29427-58-5
IUPAC Name:
methoxymethanimidamide; sulfuric acid
Synonyms:
methoxymethanimidamide; sulfuric acid
Molecular Weight (g/mol):
172.16
Molecular Formula:
C2H8N2O5S
InChi Key:
MDFRYRPNRLLJHT-UHFFFAOYSA-N
SMILES:
COC(N)=N.OS(O)(=O)=O
1
–
3
of
3
results
1
Methyl carbamimidate sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl carbamimidate sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl carbamimidate sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.