Learn More
Methyl carbamimidate sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl carbamimidate sulfate |
| CAS | 29427-58-5 |
| Molecular Formula | C2H8N2O5S |
| Quantity | 25 g |
| Synonym | O-Methylisourea Sulfate |
| InChI Formula | InChI=1S/C2H6N2O.H2O4S/c1-5-2(3)4;1-5(2,3)4/h1H3,(H3,3,4);(H2,1,2,3,4) |
| SMILES | COC(=N)N.OS(=O)(=O)O |
| IUPAC Name | methyl carbamimidate;sulfuric acid |
| Molecular Weight (g/mol) | 172.16 |
| Formula Weight | 172.0154 |