Learn More
Methyl carbamimidate sulfate, TRC
Product Code. 30468670
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468670 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30468670 Supplier Toronto Research Chemicals Supplier No. TRCM07842010G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl carbamimidate sulfate
CAS 29427-58-5
Molecular Formula C2H8N2O5S
Quantity 10 g
Synonym O-Methylisourea Sulfate
InChI Formula InChI=1S/C2H6N2O.H2O4S/c1-5-2(3)4;1-5(2,3)4/h1H3,(H3,3,4);(H2,1,2,3,4)
SMILES COC(=N)N.OS(=O)(=O)O
IUPAC Name methyl carbamimidate;sulfuric acid
Molecular Weight (g/mol) 172.16
Formula Weight 172.0154