CAS RN 293754-55-9
IUPAC Name:
N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-N-(2,2,2-trifluoroethyl)benzenesulfonamide
Synonyms:
N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-N-(2,2,2-trifluoroethyl)benzenesulfonamide
Molecular Weight (g/mol):
481.33
Molecular Formula:
C17H12F9NO3S
InChi Key:
SGIWFELWJPNFDH-UHFFFAOYSA-N
SMILES:
OC(C1=CC=C(C=C1)N(CC(F)(F)F)S(=O)(=O)C1=CC=CC=C1)(C(F)(F)F)C(F)(F)F
1
–
5
of
5
results
2
R&D Systems™ T 0901317
CAS: 293754-55-9 Molecular Formula: C17H12F9NO3S
3
T0901317, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
T0901317, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
T0901317, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.