Learn More
T0901317, TRC
Specifications
Specifications
| Chemical Name or Material | T0901317 |
| CAS | 293754-55-9 |
| Molecular Formula | C17H12F9NO3S |
| Quantity | 25 mg |
| Synonym | N-(2,2,2-Trifluoroethyl)-N-[4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl]-benzenesulfonamide, N-(2,2,2-Trifluoroethyl)-N-[4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl]benzenesulfonamide, T 1317, TO 901317 |
| InChI Formula | InChI=1S/C17H12F9NO3S/c18-14(19,20)10-27(31(29,30)13-4-2-1-3-5-13)12-8-6-11(7-9-12)15(28,16(21,22)23)17(24,25)26/h1-9,28H,10H2 |
| SMILES | COc1c(OC)cc2c(CCCNC2)c1 |
| IUPAC Name | N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-N-(2,2,2-trifluoroethyl)benzenesulfonamide |
| Molecular Weight (g/mol) | 481.33 |
| Formula Weight | 481.0394 |