accessibility menu, dialog, popup

UserName

HOBr

CAS RN 29334-16-5

IUPAC Name: (4-bromophenyl)(phenyl)methanol
Synonyms: (4-bromophenyl)(phenyl)methanol
Molecular Weight (g/mol): 263.13
Molecular Formula: C13H11BrO
InChi Key: WTIWDBNPPSHSCB-UHFFFAOYNA-N
SMILES: OC(C1=CC=CC=C1)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results