Learn More
4-Bromo-a-phenylbenzenemethanol, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-alpha-phenylbenzenemethanol |
| CAS | 29334-16-5 |
| Molecular Formula | C13 H11 Br O |
| Quantity | 50 g |
| Synonym | 4-Bromo-α-phenylbenzenemethanol, Benzhydrol, 4-bromo- (8CI), Benzohydrol, 4-bromo- (4CI), 4-Bromo-α-phenylbenzenemethanol, (4-Bromophenyl)phenylmethanol, (p-Bromophenyl)-α-phenylmethanol, (±)-(4-Bromophenyl)phenylmethanol, 4-(1-Hydroxybenzyl)bromobenzene, 4-Bromobenzhydrol, NSC 89817, p-Bromobenzhydrol, α-(4-Bromophenyl)benzenemethanol, α-Phenyl-p-bromobenzyl alcohol |
| InChI Formula | InChI=1S/C13H11BrO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9,13,15H |
| SMILES | OC(c1ccccc1)c2ccc(Br)cc2 |
| IUPAC Name | (4-bromophenyl)-phenylmethanol |
| Molecular Weight (g/mol) | 263.13 |
| Formula Weight | 261.9993 |