accessibility menu, dialog, popup

UserName

CH3OONH2HCl

CAS RN 29275-88-5

IUPAC Name: methyl 4-(aminomethyl)cyclohexane-1-carboxylate hydrochloride
Synonyms: methyl 4-(aminomethyl)cyclohexane-1-carboxylate hydrochloride
Molecular Weight (g/mol): 207.7
Molecular Formula: C9H18ClNO2
InChi Key: PZPSRTWFJCGJLP-UHFFFAOYSA-N
SMILES: Cl.COC(=O)C1CCC(CN)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results