accessibility menu, dialog, popup

UserName

ClNN

CAS RN 289905-88-0

IUPAC Name: 1-[(2-chlorophenyl)diphenylmethyl]-1H-pyrazole
Synonyms: tram-34
Molecular Weight (g/mol): 344.84
Molecular Formula: C22H17ClN2
InChi Key: KBFUQFVFYYBHBT-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(N1C=CC=N1)(C1=CC=CC=C1)C1=CC=CC=C1
Product Type 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results