Learn More
TRAM-34, TRC
Specifications
Specifications
| Chemical Name or Material | 1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole |
| CAS | 289905-88-0 |
| Molecular Formula | C22 H17 Cl N2 |
| Quantity | 25 mg |
| Synonym | 1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole |
| InChI Formula | InChI=1S/C22H17ClN2/c23-21-15-8-7-14-20(21)22(25-17-9-16-24-25,18-10-3-1-4-11-18)19-12-5-2-6-13-19/h1-17H |
| SMILES | Clc1ccccc1C(c2ccccc2)(c3ccccc3)n4cccn4 |
| IUPAC Name | 1-[(2-chlorophenyl)-diphenylmethyl]pyrazole |
| Molecular Weight (g/mol) | 344.84 |
| Formula Weight | 344.108 |