CAS RN 289042-11-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Methyl 4-(4-Fluorophenyl)-6-(1-methylethyl)-2-(methanesulfonamido)pyrimidine-5-carboxylate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-[N-methyl-N-(methylsulfonyl)amino]pyrimidine-5-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-[N-methyl-N-(methylsulfonyl)amino]pyrimidine-5-carboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.