Learn More
Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-[N-methyl-N-(methylsulfonyl)amino]pyrimidine-5-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-[N-methyl-N-(methylsulfonyl)amino]pyrimidine-5-carboxylate |
| CAS | 289042-11-1 |
| Molecular Formula | C17H20FN3O4S |
| Quantity | 100 mg |
| Synonym | 5-Pyrimidinecarboxylic acid','4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-','methyl ester, 4-(4-Fluorophenyl)-6-isopropyl-2-[(methanesulfonyl)methylamino]pyrimidine-5-carboxylic acid methyl ester, Methyl 4-(4-fluorophenyl)-6-isopropyl-2-[N-methyl-N-(methylsulfonyl)amino]pyrimidine-5-carboxylate, Methyl 4-(4-Fluorophenyl)-6-(1-methylethyl)-2-(methanesulfonamido)pyrimidine-5-carboxylate |
| InChI Formula | InChI=1S/C17H20FN3O4S/c1-10(2)14-13(16(22)25-4)15(11-6-8-12(18)9-7-11)20-17(19-14)21(3)26(5,23)24/h6-10H,1-5H3 |
| SMILES | COC(=O)c1c(nc(nc1c2ccc(F)cc2)N(C)S(=O)(=O)C)C(C)C |
| IUPAC Name | methyl 4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidine-5-carboxylate |
| Molecular Weight (g/mol) | 381.42 |
| Formula Weight | 381.1159 |