CAS RN 287476-11-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Melamine-15N3, TRC
CAS: 287476-11-3 Molecular Formula: C3 H6 15N3 N3 Molecular Weight (g/mol): 129.1 Synonym: Melamine triamine 15N3,Melamine Triamine-15N3,1,3,5-Triazine-2,4,6-tri(amine-15N) (9CI, ACI) IUPAC Name: 1,3,5-triazine-2,4,6-triamine SMILES: [15NH2]c1nc([15NH2])nc([15NH2])n1