Learn More
Melamine-15N3, TRC
Product Code. 30011172
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011172 1 mg 1mg
30006590 10 mg 10mg
30004050 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011172 Supplier Toronto Research Chemicals Supplier No. TRCM2087031MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 287476-11-3
Molecular Formula C3 H6 15N3 N3
Molecular Weight (g/mol) 129.1
Synonym Melamine triamine 15N3, Melamine Triamine-15N3, 1,3,5-Triazine-2,4,6-tri(amine-15N) (9CI, ACI)
IUPAC Name 1,3,5-triazine-2,4,6-triamine
SMILES [15NH2]c1nc([15NH2])nc([15NH2])n1
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)/i4+1,5+1,6+1
Formula Weight 129.0565
Percent Purity >95%
Chemical Name or Material Melamine-15N3
Grade HPLC