accessibility menu, dialog, popup

UserName

CH3CH3OOOH3CCH3

CAS RN 28434-01-7

IUPAC Name: (5-benzylfuran-3-yl)methyl 2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate
Synonyms: for-syn chryson benzyfuroline resmetrina ari-B penick 1382 penncapthrin SB pennick 1382 bioresmethrin (d trans isomer) synthrin
Molecular Weight (g/mol): 338.45
Molecular Formula: C22H26O3
InChi Key: VEMKTZHHVJILDY-UHFFFAOYNA-N
SMILES: CC(C)=CC1C(C(=O)OCC2=COC(CC3=CC=CC=C3)=C2)C1(C)C
Concentration 
  • (3)
Solvent or Matrix 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results