CAS RN 23982-76-5
CAS RN 23982-76-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ 10-Cl-BBQ
Potent aryl hydrocarbon receptor (AhR) agonist; orally bioavailable