accessibility menu, dialog, popup

UserName

CAS RN 22902-84-7

FFFClOOH

CAS RN 22902-84-7

IUPAC Name: 2-[2-chloro-3-(trifluoromethyl)phenyl]acetic acid
Synonyms: [2-chloro-3-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: JZEWYZAIWWNAPZ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(Cl)C(=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results