CAS RN 22881-33-0
CAS RN 22881-33-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[(p-chlorobenzyl)amino]-pyridine, TRC
CAS: 22881-33-0 Molecular Formula: C12 H11 Cl N2 Molecular Weight (g/mol): 218.68 Synonym: N-[(4-chlorophenyl)methyl]-2-pyridinamine,2-[(p-Chlorobenzyl)amino]pyridine,NSC 118980,Chloropyramine Impurity IUPAC Name: N-[(4-chlorophenyl)methyl]pyridin-2-amine SMILES: Clc1ccc(CNc2ccccn2)cc1