missing translation for 'onlineSavingsMsg'
Learn More

2-[(p-chlorobenzyl)amino]-pyridine, TRC

Product Code. 30137435
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137435 5 g 5g
30012614 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137435 Supplier Toronto Research Chemicals Supplier No. TRCC3647455G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22881-33-0
Molecular Formula C12 H11 Cl N2
Molecular Weight (g/mol) 218.68
Synonym N-[(4-chlorophenyl)methyl]-2-pyridinamine, 2-[(p-Chlorobenzyl)amino]pyridine, NSC 118980, Chloropyramine Impurity
IUPAC Name N-[(4-chlorophenyl)methyl]pyridin-2-amine
SMILES Clc1ccc(CNc2ccccn2)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H11ClN2/c13-11-6-4-10(5-7-11)9-15-12-3-1-2-8-14-12/h1-8H,9H2,(H,14,15)
Formula Weight 218.0611
Chemical Name or Material p-Chlorobenzylaminopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.