accessibility menu, dialog, popup

UserName

HOOFCl

CAS RN 2252-50-8

IUPAC Name: 2-chloro-5-fluorobenzoic acid
Synonyms: 2-chloro-5-fluorobenzoic acid
Molecular Weight (g/mol): 174.56
Molecular Formula: C7H4ClFO2
InChi Key: MIZKCMSSYVUZKD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results