missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorobenzoic acid, 97%, Thermo Scientific™

Product Code. 11435810
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435810 25 g 25g
11415810 1 g 1g
11425810 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435810 Supplier Thermo Scientific Alfa Aesar Supplier No. H26462.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 2252-50-8
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.56
MDL Number MFCD00156967
InChI Key MIZKCMSSYVUZKD-UHFFFAOYSA-N
Synonym 5-fluoro-2-chlorobenzoic acid, 2-chloro-5-fluorobenzoicacid, 2-chloro-5-fluoro-benzoic acid, 2-chloro5-fluorobenzoic acid, 2-chloro-5-fluoro benzoic acid, pubchem1371, acmc-209fx4, ksc201s8l, rarechem al bo 2229
PubChem CID 302888
IUPAC Name 2-chloro-5-fluorobenzoic acid
SMILES OC(=O)C1=CC(F)=CC=C1Cl

Spezifikation

Melting Point 147°C to 150°C
Quantity 25 g
Formula Weight 174.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-fluorobenzoic acid
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.