accessibility menu, dialog, popup

UserName

NH2OFF

CAS RN 22236-04-0

IUPAC Name: 2-(difluoromethoxy)aniline
Synonyms: 2-(difluoromethoxy)aniline
Molecular Weight (g/mol): 159.14
Molecular Formula: C7H7F2NO
InChi Key: CGNAIUUCOVWLLL-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1OC(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results