missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 11385299
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11385299 5 g 5g
11375299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385299 Supplier Thermo Scientific Alfa Aesar Supplier No. B20255.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22236-04-0
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.14
MDL Number MFCD00190104
InChI Key CGNAIUUCOVWLLL-UHFFFAOYSA-N
Synonym 2-difluoromethoxy aniline, o-difluoromethoxyaniline, 2-difluoromethoxy-phenylamine, benzenamine, 2-difluoromethoxy, 2-difluoromethoxy phenylamine, pubchem8465, acmc-1cll9, akos bb184, asischem y89474, ksc494k1l
PubChem CID 934620
IUPAC Name 2-(difluoromethoxy)aniline
SMILES NC1=CC=CC=C1OC(F)F

Specifications

Density 1.265
Boiling Point 205°C
Flash Point 92°C (197°F)
Odor Characteristic
Refractive Index 1.504
Quantity 5 g
UN Number UN2810
Beilstein 2718520
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethoxy)aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.