accessibility menu, dialog, popup

UserName

CAS RN 21679-14-1

N(R)N(S)O(S)(R)NNOHNH2HOHOF

CAS RN 21679-14-1

IUPAC Name: (2R,3S,4S,5R)-2-(6-amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: fludarabine
Molecular Weight (g/mol): 285.24
Molecular Formula: C10H12FN5O4
InChi Key: HBUBKKRHXORPQB-FJFJXFQQSA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O)C2=NC(F)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results