accessibility menu, dialog, popup

UserName

CAS RN 21678-37-5

CH3H3CONCH3H3C

CAS RN 21678-37-5

IUPAC Name: N,N,2-trimethylpropanamide
Synonyms: N,N,2-trimethylpropanamide
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: GXMIHVHJTLPVKL-UHFFFAOYSA-N
SMILES: CC(C)C(=O)N(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results