accessibility menu, dialog, popup

UserName

CAS RN 2114-00-3

CH3BrO

CAS RN 2114-00-3

IUPAC Name: 2-bromo-1-phenylpropan-1-one
Synonyms: 1-propanone, 2-bromo-1-phenyl-
Molecular Weight (g/mol): 213.07
Molecular Formula: C9H9BrO
InChi Key: WPDWOCRJBPXJFM-UHFFFAOYNA-N
SMILES: CC(Br)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)