Learn More
Alpha-Bromopropiophenone, TRC
Specifications
Specifications
| Chemical Name or Material | alpha-Bromopropiophenone |
| CAS | 2114-00-3 |
| Molecular Formula | C9H9BrO |
| Quantity | 25 g |
| Synonym | 2-Bromo-1-phenyl-1-propanone, 1-Benzoyl-1-bromoethane, 1-Bromoethyl Phenyl Ketone, 2-Bromo-1-phenyl-1-propanone, dl-alpha-Bromopropiophenone, NSC 89689 |
| InChI Formula | InChI=1S/C9H9BrO/c1-7(10)9(11)8-5-3-2-4-6-8/h2-7H,1H3 |
| SMILES | CC(Br)C(=O)c1ccccc1 |
| IUPAC Name | 2-bromo-1-phenylpropan-1-one |
| Molecular Weight (g/mol) | 213.07 |
| Formula Weight | 211.9837 |