Learn More
Gentiopicroside, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Gentiopicroside |
| CAS | 20831-76-9 |
| Molecular Formula | C16 H20 O9 |
| Quantity | 25 mg |
| Synonym | 1H,3H-Pyrano[3,4-c]pyran-1-one, 5-ethenyl-6-(β-D-glucopyranosyloxy)-5,6-dihydro-, (5R,6S)-, 1H,3H-Pyrano[3,4-c]pyran-1-one, 5-ethenyl-6-(β-D-glucopyranosyloxy)-5,6-dihydro-, (5R-trans)-, Gentiopicrin (6CI), Gentiopicroside (7CI,8CI), (5R,6S)-5-Ethenyl-6-(β-D-glucopyranosyloxy)-5,6-dihydro-1H,3H-pyrano[3,4-c]pyran-1-one, NSC 606402 |
| InChI Formula | InChI=1S/C16H20O9/c1-2-7-8-3-4-22-14(21)9(8)6-23-15(7)25-16-13(20)12(19)11(18)10(5-17)24-16/h2-3,6-7,10-13,15-20H,1,4-5H2/t7-,10-,11-,12+,13-,15+,16+/m1/s1 |
| SMILES | OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC=C3[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC Name | (3S,4R)-4-ethenyl-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,6-dihydro-3H-pyrano[3,4-c]pyran-8-one |
| Molecular Weight (g/mol) | 356.32 |
| Formula Weight | 356.1107 |