accessibility menu, dialog, popup

UserName

CAS RN 2052-15-5

H3COOCH3O

CAS RN 2052-15-5

IUPAC Name: butyl 4-oxopentanoate
Synonyms: butyl laevulinate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: ISBWNEKJSSLXOD-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CCC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results