accessibility menu, dialog, popup

UserName

NH2OClNOO

CAS RN 2011-66-7

IUPAC Name: 2-(2-chlorobenzoyl)-4-nitroaniline
Synonyms: 2-(2-chlorobenzoyl)-4-nitroaniline
Molecular Weight (g/mol): 276.68
Molecular Formula: C13H9ClN2O3
InChi Key: GRDGBWVSVMLKBV-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1C(=O)C1=CC=CC=C1Cl)[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results