accessibility menu, dialog, popup

UserName

CAS RN 19393-96-5

ClClBrCl

CAS RN 19393-96-5

IUPAC Name: 2-bromo-1,3,5-trichlorobenzene
Synonyms: 2-bromo-1,3,5-trichlorobenzene
Molecular Weight (g/mol): 260.34
Molecular Formula: C6H2BrCl3
InChi Key: BAPPAFMEUDJAQI-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(Br)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results